Tetracycline

Supplier: Molekula

32100101-25G
MOLE32100101-25GEA 70.1 EUR
MOLE32100101-25G
Tetracycline
Tetracycline
Formula: C₂₂H₂₄N₂O₈
MW: 444.43 g/mol
MDL Number: MFCD00151232
CAS Number: 60-54-8
EINECS: 200-481-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR