Dichloroacetic acid (DCA)

Supplier: Molekula
Danger

Synonyms: DCA Deblock, DCA

32742152-1KG
MOLE32742152-1KGEA 140 EUR
MOLE32742152-1KG
Dichloroacetic acid (DCA)
Dichloroacetic acid (DCA)
Formula: C₂H₂Cl₂O₂
MW: 128.94 g/mol
Boiling Pt: 194
Melting Pt: ∼–4 °C
Density: 1.563 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004223
CAS Number: 79-43-6
EINECS: 201-207-0
UN: 1764
ADR: 8 II
Merck Index: 13,03075

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR