(R)-(+)-β-Methylphenethylamine

Supplier: Molekula

Synonyms: (R)-(+)-beta-Methylphenethylamine, (R)-(+)-2-Phenyl-1-propylamine

37493559-1G
MOLE37493559-1GEA 0 EUR
MOLE37493559-1G
(R)-(+)-β-Methylphenethylamine
(R)-(+)-β-Methylphenethylamine
Formula: C₉H₁₃N
MDL Number: MFCD00216740
CAS Number: 28163-64-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR