(4-Bromophenyl)acetonitrile

Supplier: Molekula

Synonyms: 4-Bromophenylacetonitrile, 4-Bromobenzyl cyanide

38110135-100G
MOLE38110135-100GEA 0 EUR
MOLE38110135-100G
(4-Bromophenyl)acetonitrile
(4-Bromophenyl)acetonitrile
Formula: C₈H₆BrN
MW: 196,05 g/mol
Boiling Pt: 286 °C (1013 hPa)
Melting Pt: 47…49 °C
MDL Number: MFCD00001916
CAS Number: 16532-79-9
EINECS: 240-602-2
UN: 3449
ADR: 6.1,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR