3-(Dimethylamino)benzoic acid

Supplier: Molekula

38169682-100G
MOLE38169682-100GEA 0 EUR
MOLE38169682-100G
3-(Dimethylamino)benzoic acid
3-(Dimethylamino)benzoic acid
Formula: C₉H₁₁NO₂
Boiling Pt: 298 °C (1013 hPa)
Melting Pt: 149…152 °C
Density: 1,26 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002497
CAS Number: 99-64-9
EINECS: 202-775-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR