1-Bromo-4-propylbenzene

Supplier: Molekula

38940636-25G
MOLE38940636-25GEA 0 EUR
MOLE38940636-25G
1-Bromo-4-propylbenzene
1-Bromo-4-propylbenzene
Formula: C₉H₁₁Br
Boiling Pt: 224 °C (1013 hPa)
Melting Pt: –41,5 °C
Density: 1,283 g/cm³ (20 °C)
MDL Number: MFCD00012456
CAS Number: 588-93-2
EINECS: 454-790-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR