Potassium tellurite hydrate 98

Supplier: Molekula
Danger

Synonyms: Potassium tellurite(IV) hydrate, Potassium tellurium trioxide hydrate, di-Potassium tellurium trioxide hydrate, Potassium tellurium (IV) oxide hydrate, Potassium tellurite monohydrate

40505087-100G
MOLE40505087-100GEA 633 EUR
MOLE40505087-100G
Potassium tellurite hydrate 98
di-Potassium tellurium trioxide hydrate
Formula: K₂O₃Te·H₂O
MW: 271.81 g/mol
MDL Number: MFCD00149923
CAS Number: 123333-66-4
EINECS: 232-213-1
UN: 3284
ADR: 6.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR