Dimethyl oxalate

Supplier: Molekula
Warning

Synonyms: Oxalic acid dimethyl ester

41260542-500G
MOLE41260542-500GEA 0 EUR
MOLE41260542-500G
Dimethyl oxalate
Dimethyl oxalate
Formula: C₄H₆O₄
MW: 118.09 g/mol
Boiling Pt: 163
Melting Pt: 54
Density: 1.422 g/cm³ (15 °C)
Flash Pt: 62
Storage Temperature: Ambient
MDL Number: MFCD00008442
CAS Number: 553-90-2
EINECS: 209-053-6
UN: 3261
Merck Index: 12,06181

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR