α,2-Dibromotoluene

Supplier: Molekula
Danger

Synonyms: 2-Bromobenzyl bromide

45245256-25G
MOLE45245256-25GEA 107 EUR
MOLE45245256-25G
α,2-Dibromotoluene
α,2-Dibromotoluene
Formula: C₇H₆Br₂
MW: 249.93 g/mol
Boiling Pt: 253
Melting Pt: 34
Density: 1.881 g/cm³ (20 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00000173
CAS Number: 3433-80-5
EINECS: 222-334-8
UN: 2293
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR