D-(-)-Luciferin

Supplier: Molekula

45944318-25MG
MOLE45944318-25MGEA 0 EUR
MOLE45944318-25MG
D-(-)-Luciferin
D(-)-Luciferin
Formula: C₁₁H₈N₂O₃S₂
MDL Number: MFCD00042929
CAS Number: 2591-17-5
EINECS: 219-981-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR