D-(+)-Allose

Supplier: Molekula

50241498-250MG
MOLE50241498-250MGEA 143 EUR
MOLE50241498-250MG
D-(+)-Allose
D(+)-Allose
Formula: C₆H₁₂O₆
MDL Number: MFCD00135833
CAS Number: 2595-97-3
EINECS: 219-994-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR