2-Nitrobiphenyl

Supplier: Molekula

52908098-25G
MOLE52908098-25GEA 143 EUR
MOLE52908098-25G
2-Nitrobiphenyl
2-Nitrobiphenyl
Formula: C₁₂H₉NO₂
Boiling Pt: 320 °C (1013 hPa)
Melting Pt: 36 °C
Density: 1,44 g/cm³ (20 °C)
MDL Number: MFCD00007126
CAS Number: 86-00-0
EINECS: 201-646-8
Merck Index: 12,06689

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR