4-Hydroxy-3-methoxybenzyl alcohol

Supplier: Molekula

63880340-100G
MOLE63880340-100GEA 0 EUR
MOLE63880340-100G
4-Hydroxy-3-methoxybenzyl alcohol
4-Hydroxy-3-methoxybenzyl alcohol
Formula: C₈H₁₀O₃
Boiling Pt: 293 °C (1013 hPa)
Melting Pt: 113 °C
MDL Number: MFCD00004659
CAS Number: 498-00-0
EINECS: 207-852-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR