2,3-Dichloroaniline

Supplier: Molekula

68153911-500G
MOLE68153911-500GEA 0 EUR
MOLE68153911-500G
2,3-Dichloroaniline
2,3-Dichloroaniline
Formula: C₆H₅Cl₂N
Boiling Pt: 252 °C (1013 hPa)
Melting Pt: 23…24 °C
Density: 1,383 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007657
CAS Number: 608-27-5
EINECS: 210-157-9
UN: 3442
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR