9-Anthracenecarboxylic acid

Supplier: Molekula

Synonyms: 9-Anthroic acid, Anthracene-9-carboxylic acid, 9-Anthracenecarboxylic acid, Anthracene-9-carboxylic acid, 9-Carboxyanthracene

72853037-25G
MOLE72853037-25GEA 70.1 EUR
MOLE72853037-25G
9-Anthracenecarboxylic acid
9-Anthracenecarboxylic acid
Formula: C₁₅H₁₀O₂
MW: 222.24 g/mol
Boiling Pt: 411
Melting Pt: 214…217 °C
Density: 1.397 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001257
CAS Number: 723-62-6
EINECS: 211-964-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR