Hydroxylamine-O-sulphonic acid

Supplier: Molekula

77461217-100G
MOLE77461217-100GEA 141 EUR
MOLE77461217-100G
Hydroxylamine-O-sulphonic acid
Hydroxylamine-O-sulphonic acid
Formula: H₃NO₄S
MDL Number: MFCD00011604
CAS Number: 2950-43-8
EINECS: 220-971-6
UN: 3260
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR