2-Aminothiophenol

Supplier: Molekula

Synonyms: 2-Aminophenyl mercaptan, 2-Aminobenzenethiol, 2-Mercaptoaniline

85307137-500G
MOLE85307137-500GEA 408 EUR
MOLE85307137-500G
2-Aminothiophenol
2-Aminothiophenol
Formula: C₆H₇NS
MW: 125.19 g/mol
Boiling Pt: 234
Melting Pt: 26
Density: 1.17 g/cm³ (20 °C)
Flash Pt: 79
Storage Temperature: Refrigerator
MDL Number: MFCD00007702
CAS Number: 137-07-5
EINECS: 205-277-3
UN: 3267
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR