Phenothiazine

Supplier: Molekula
Warning

90028302
MOLE90028302EA 0 EUR
MOLE90028302
Phenothiazine
Phenothiazine
Formula: C₁₂H₉NS
Boiling Pt: 371 °C (1013 hPa)
Melting Pt: 182…185 °C
Density: 1,34 g/cm³ (20 °C)
Flash Pt: 202 °C
MDL Number: MFCD00005015
CAS Number: 92-84-2
EINECS: 202-196-5
Merck Index: 12,07404

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR