Creatinine

Supplier: Molekula

M25285847
MOLEM25285847EA 0 EUR
MOLEM25285847
Creatinine
Creatinine
Formula: C₄H₇N₃O
MDL Number: MFCD00059730
CAS Number: 60-27-5
EINECS: 200-466-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR