2-Phenylaniline

Supplier: Molekula
Warning

M26380604
MOLEM26380604EA 0 EUR
MOLEM26380604
2-Phenylaniline
2-Phenylaniline
Formula: C₁₂H₁₁N
Boiling Pt: 299 °C (1013 hPa)
Melting Pt: 42…44 °C
Storage Temperature: Ambient
MDL Number: MFCD00007701
CAS Number: 90-41-5
EINECS: 201-990-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR