Methyl green zinc chloride salt

Supplier: Molekula
Warning

Synonyms: CI 42590, Ethyl Green, Methyl green zinc chloride double salt, Methyl Green

M31140436
MOLEM31140436EA 0 EUR
MOLEM31140436
Methyl green zinc chloride salt
Methyl Green zinc chloride salt
Formula: C₂₇H₃₅BrClN₃·xZnCl₂
Storage Temperature: Ambient
MDL Number: MFCD00151094
CAS Number: 7114-03-6
EINECS: 230-415-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR