Brilliant green

Supplier: Molekula
Warning

M31475505
MOLEM31475505EA 0 EUR
MOLEM31475505
Brilliant green
Brilliant Green
Formula: C₂₇H₃₄N₂O₄S
Storage Temperature: Ambient
MDL Number: MFCD00011880
CAS Number: 633-03-4
EINECS: 211-190-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR