Syringic acid

Supplier: Molekula

Synonyms: 4-Hydroxy-3,5-dimethoxybenzoic acid

M33897973
MOLEM33897973EA 0 EUR
MOLEM33897973
Syringic acid
Syringic acid
Formula: C₉H₁₀O₅
Boiling Pt: 351 °C (1013 hPa)
Melting Pt: 204…207 °C
Storage Temperature: Ambient
MDL Number: MFCD00002552
CAS Number: 530-57-4
EINECS: 208-486-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR