Aluminium trifluoromethanesulphonate

Supplier: Molekula

M38589392
MOLEM38589392EA 0 EUR
MOLEM38589392
Aluminium trifluoromethanesulphonate
Aluminium trifluoromethanesulphonate
Formula: C₃AlF₉O₉S₃
MDL Number: MFCD00143596
CAS Number: 74974-61-1
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR