Ethyl-3,4-dihydroxybenzoate

Supplier: Molekula

M43205547
MOLEM43205547EA 0 EUR
MOLEM43205547
Ethyl-3,4-dihydroxybenzoate
Ethyl 3,4-dihydroxybenzoate
Formula: C₉H₁₀O₄
Melting Pt: 134 °C
MDL Number: MFCD00002199
CAS Number: 3943-89-3
EINECS: 223-529-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR