2,6-Dihydroxynaphthalene

Supplier: Molekula
Warning

Synonyms: Naphthalene-2,6-diol

M45946669
MOLEM45946669EA 0 EUR
MOLEM45946669
2,6-Dihydroxynaphthalene
2,6-Dihydroxynaphthalene
Formula: C₁₀H₈O₂
MW: 160.17 g/mol
Boiling Pt: 375
Melting Pt: 220
Density: 1.33 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004082
CAS Number: 581-43-1
EINECS: 209-465-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR