Acetylcholine bromide

Supplier: Molekula

Synonyms: 2-Acetoxy-N,N,N-trimethylethanaminium bromide, (2-Acetoxyethyl)trimethylammonium bromide

M54740269
MOLEM54740269EA 0 EUR
MOLEM54740269
Acetylcholine bromide
Acetylcholine bromide
Formula: H₃CC(O)O(CH₂)₂N(CH₃)₃Br
MW: 226.11 g/mol
Melting Pt: 142…145 °C
Density: 1.467 g/cm³ (20 °C)
Storage Temperature: Freezer
MDL Number: MFCD00011814
CAS Number: 66-23-9
EINECS: 200-622-4
Merck Index: 13,00088

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR