2'-Bromoacetophenone

Supplier: Molekula
Warning

Synonyms: o-Bromoacetophenone

M57640263
MOLEM57640263EA 0 EUR
MOLEM57640263
2'-Bromoacetophenone
2'-Bromoacetophenone
Formula: C₈H₇BrO
MW: 199,05 g/mol
Boiling Pt: 249 °C (1013 hPa)
Melting Pt: 0 °C
Density: 1,476 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000067
CAS Number: 2142-69-0
EINECS: 218-398-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR