3-Nitrobenzaldehyde

Supplier: Molekula

M63370135
MOLEM63370135EA 0 EUR
MOLEM63370135
3-Nitrobenzaldehyde
3-Nitrobenzaldehyde
Formula: C₇H₅NO₃
MW: 151.12 g/mol
Boiling Pt: 232
Melting Pt: 58
Density: 1.481 g/cm³ (0 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007249
CAS Number: 99-61-6
EINECS: 202-772-6
Merck Index: 12,06684

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR