Thiosalicylic acid

Supplier: Molekula
Warning

Synonyms: 2-Thiobenzoic acid, 2-Mercaptobenzoic acid

M63776863
MOLEM63776863EA 0 EUR
MOLEM63776863
Thiosalicylic acid
Thiosalicylic acid
Formula: C₇H₆O₂S
Boiling Pt: 230 °C (0,02 mmHg)
Melting Pt: 165…166 °C
Density: 1,489 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004836
CAS Number: 147-93-3
EINECS: 205-704-3
UN: 3335
ADR: 9,
Merck Index: 12,09498

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR