n-Nonadecane

Supplier: Molekula

M67720907
MOLEM67720907EA 0 EUR
MOLEM67720907
n-Nonadecane
n-Nonadecane
Formula: H₃C(CH₂)₁₇CH₃
Boiling Pt: 329,7 °C (1013 hPa)
Melting Pt: 32,5 °C
Density: 0,7845 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009012
CAS Number: 629-92-5
EINECS: 211-116-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR