2-Methylbenzoxazole

Supplier: Molekula

M68142081
MOLEM68142081EA 0 EUR
MOLEM68142081
2-Methylbenzoxazole
2-Methylbenzoxazole
Formula: C₈H₇NO
Boiling Pt: 200 °C (1013 hPa)
Density: 1,12 g/cm³ (20 °C)
MDL Number: MFCD00005771
CAS Number: 95-21-6
EINECS: 202-399-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR