Fumaroyl chloride

Supplier: Molekula

Synonyms: Fumaryl chloride

M69920700
MOLEM69920700EA 0 EUR
MOLEM69920700
Fumaroyl chloride
Fumaroyl chloride
Formula: C₄H₂Cl₂O₂
MW: 152.96 g/mol
Boiling Pt: 161
Melting Pt: –2 °C
Density: 1.41 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000733
CAS Number: 627-63-4
EINECS: 211-005-4
UN: 1780
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR