D-α-Phenylalanine

Supplier: Molekula

Synonyms: D-Phenylalanine, H-D-Phe-OH, 3-Phenyl-D-alanine, 3-Phenyl-D(+)-alanine

M76521605
MOLEM76521605EA 0 EUR
MOLEM76521605
D-α-Phenylalanine
D-α-Phenylalanine
Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Boiling Pt: 295
MDL Number: MFCD00004270
CAS Number: 673-06-3
EINECS: 211-603-5
Merck Index: 12,07425

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR