4-Methyl-1,10-phenanthroline

Supplier: Molekula

M76544185
MOLEM76544185EA 0 EUR
MOLEM76544185
4-Methyl-1,10-phenanthroline
4-Methyl-1,10-phenanthroline
Formula: C₁₃H₁₀N₂
MW: 194.24 g/mol
MDL Number: MFCD00004978
CAS Number: 31301-28-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR