Cresol red

Supplier: Molekula

M79737710
MOLEM79737710EA 0 EUR
MOLEM79737710
Cresol red
Cresol red
Formula: C₂₁H₁₈O₅S
Storage Temperature: Ambient
MDL Number: MFCD00005878
CAS Number: 1733-12-6
EINECS: 217-064-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR