tert-Butyldimethylsilyl trifluoromethanesulphonate

Supplier: Molekula

M84017698
MOLEM84017698EA 0 EUR
MOLEM84017698
tert-Butyldimethylsilyl trifluoromethanesulphonate
tert-Butyldimethylsilyl trifluoromethanesulphonate
Formula: C₇H₁₅F₃O₃SSi
MDL Number: MFCD00000405
CAS Number: 69739-34-0
EINECS: 274-102-0
UN: 2920
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR