4'-(Trifluoromethyl)acetophenone

Supplier: Molekula
Warning

Synonyms: 1-[4-(Trifluoromethyl)phenyl]ethan-1-one, 4-(Trifluoromethyl)acetophenone, p-(Trifluoromethyl)acetophenone

M84087027
MOLEM84087027EA 0 EUR
MOLEM84087027
4'-(Trifluoromethyl)acetophenone
4'-(Trifluoromethyl)acetophenone
Formula: C₉H₇F₃O
Boiling Pt: 205 °C (1013 hPa)
Melting Pt: 26…29 °C
Density: 0,924 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000401
CAS Number: 709-63-7
EINECS: 211-913-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR