Sebacic acid

Supplier: Molekula

Synonyms: Decanedioic acid, 1,8-Octanedicarboxylic acid, 1,10-Decanedioic acid

M86335635
MOLEM86335635EA 0 EUR
MOLEM86335635
Sebacic acid
Sebacic acid
Formula: C₁₀H₁₈O₄
MW: 202.25 g/mol
Boiling Pt: 374
Melting Pt: 127…131 °C
Density: 1.21 g/cm³ (20 °C)
Flash Pt: 221
Storage Temperature: Ambient
MDL Number: MFCD00004440
CAS Number: 111-20-6
EINECS: 203-845-5
REACH: 01-2119519212-52
Merck Index: 12,08558

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR