7-Methoxyindole

Supplier: Molekula
Warning

Synonyms: 7-Methoxy-1H-indole, Indol-7-yl methyl ether

M89429300
MOLEM89429300EA 0 EUR
MOLEM89429300
7-Methoxyindole
7-Methoxyindole
Formula: C₉H₉NO
MW: 147,18 g/mol
Boiling Pt: 298 °C (1013 hPa)
Density: 1,126 g/cm³ (25 °C)
Storage Temperature: Freezer
MDL Number: MFCD00047203
CAS Number: 3189-22-8
EINECS: 221-690-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR