Nile blue A ≥75 % (dye content), Sigma-Aldrich®

Supplier: SIGMA ALDRICH MICROSCOPY

N5632-25G
N5632-25GEA 91 EUR
N5632-25G
Nile blue A ≥75 % (dye content), Sigma-Aldrich®
Nile blue A

Nile Blue A, Dye content ≥75 % (Basic Blue 12; Nile blue sulfate)

Formula: C₄₀H₄₀N₆O₆S
Melting Pt: >300 °C (dec., lit.)
Storage Temperature: Ambient
MDL Number: MFCD00064529
CAS Number: 3625-57-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c1e9a300af6fcdad8689d11e210d54d4e80d7765&vid=bcfa12b44332210932aeadb7cb05764c10a14512&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR