Anti-DNA-PK (Ab-2) (18-2) Mouse Monoclonal Antibody [clone: 42053]

Supplier: Merck Millipore (Oncogene)
NA57-100
NA57-100EA 722 EUR
NA57-100
Anti-DNA-PK (Ab-2) (18-2) Mouse Monoclonal Antibody [clone: 42053]
Antibodies
Type: Primary
Antigen: DNA-PK (Ab-2) (18-2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR