Anti-BrdU (Ab-3) (Mobu-1) Mouse Monoclonal Antibody

Supplier: Merck Millipore (Oncogene)
NA61-100
NA61-100EA 704 EUR
NA61-100
Anti-BrdU (Ab-3) (Mobu-1) Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: BrdU (Ab-3) (Mobu-1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Pan
Order Now

SPECIFICATIONS

Antigen symbol BRDU
Environmentally Preferable
Antibody type Primary
Clonality Monoclonal
Conjugation Unconjugated
Reactivity Multiple
Host Mouse
Isotype
ImmunoChemistry Yes
ImmunoFluorescence Yes
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR