Non-phthalate PVC tubing, Nalgene™
Supplier: Thermo Fisher Scientific
8701-0010 8701-0020 8701-9020 8701-0030 8701-9030 8701-0060 8701-4060 8701-0070 8701-0080 8701-0090 8701-9090 8701-0100 8701-0110 8701-0120 8701-4120 8701-0130 8701-0140 8701-4140 8701-0160 8701-0180 8701-0190 8701-0200 8701-0260 8701-0310 8701-0350
NALG8701-0010EA 0 EUR
NALG8701-0010 NALG8701-0020 NALG8701-9020 NALG8701-0030 NALG8701-9030 NALG8701-0060 NALG8701-4060 NALG8701-0070 NALG8701-0080 NALG8701-0090 NALG8701-9090 NALG8701-0100 NALG8701-0110 NALG8701-0120 NALG8701-4120 NALG8701-0130 NALG8701-0140 NALG8701-4140 NALG8701-0160 NALG8701-0180 NALG8701-0190 NALG8701-0200 NALG8701-0260 NALG8701-0310 NALG8701-0350
Non-phthalate PVC tubing, Nalgene™
Tubing Transfer Tubing
Nalgene lab-grade, autoclavable clear tubing sets the standard for purity and quality, made from highest-grade resins, free from ortho-phthalate plasticisers.
  • Compare to Tygon® E-3603 tubing
  • Marked with Int.Ø and regulatory compliance as well as at 305 mm intervals for easy identification and measurement
  • Durometer hardness is shore A, 60
Nalgene™ Non-phthalate PVC tubing is manufactured from the highest-grade resin materials and performs like classic Nalgene™ 180 tubing but is free from orthophthalate DEHP plasticisers. Resists hardening andMore Product Information