Blender bags, Whirl-Pak®
Supplier: NASCO FORT ATKINSON
B01239WA B01421WA B01474WA
NASCB01239WAEA 866 EUR
NASCB01239WA NASCB01421WA NASCB01474WA
Blender bags, Whirl-Pak®
Bags Blender Bags
Blended, virgin polyethylene film provides exceptional clarity, and superior strength for use in homogenizer. HACCP, EPA, USDA, FDA approved for food contact.
  • Plus 1 sterilisation, post-manufacturing sterilisation: Increased sample integrity, perforated closure to maintain sterility
  • Continuous perimeter seal: Maximum protection against leaks
This bag is specially designed for use with homogenizer blenders. It is made from extra-heavy polyethylene and has special features to make sample testing easier and more efficient.
Shelf life fromMore Product Information