Boiling baths, analogue, clifton, NE1B Series
Supplier: NICKEL ELECTRO
NE1B-14 NE1B-28
NE1B-14EA 2460 EUR
NE1B-14 NE1B-28
Boiling baths, analogue, clifton, NE1B Series
Baths Boiling Baths
Designed for applications that require continuous boiling. Baths have a corrosion resistant stainless steel working environment, with heater and safety cut out concealed underneath, all housed in a durable, chemical resistant, powder coated paint finished outer case.
  • Integral constant level device for connection to water supply combats water evaporation and maintains boiling
  • Include 'low' height removable stainless steel shelf to increase working volume
  • Drain outlet on 28 litre model for convenient bath emptying
  • Boiling can be regulated from vigorous to a
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable