Crucibles, stainless steel
Supplier: NICKEL ELECTRO
123 124 126
NICK123EA 21.2 EUR
NICK123 NICK124 NICK126
Crucibles, stainless steel
Crucibles
These stainless steel 304 crucibles with bright finish are much harder and last longer than porcelain types.
  • Non-magnetic
  • Shatterproof
Ordering information: Lids need to be ordered separately.