Heavy duty hoses for airhoods
Supplier: Honeywell Safety
A161253
NORHA161253EA 696 EUR
NORHA161253
Heavy duty hoses for airhoods
Tubing Vacuum/Pressure Tubing
Black heavy duty breathing hoses for use in connection with light and heavy duty airhoods.
  • Anti-static
  • With quick connection double action safety male and female couplings
  • Available in several lengths
Certifications: EN 139