Anti-IRE1 alpha (phospho-Ser724) Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB100-2323
NOVUNB100-2323EA 734 EUR
NOVUNB100-2323
Anti-IRE1 alpha (phospho-Ser724) Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal IRE1 alpha [p Ser724] Antibody. Tested Applications: Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human, Mouse, Rat, Primate (-).

Type: Primary
Antigen: IRE1 alpha
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR