Anti-Speedy/Ringo Rabbit Polyclonal Antibody (DyLight® 550)

Supplier: Novus Biologicals
NB100-2521R
NOVUNB100-2521REA 722 EUR
NOVUNB100-2521R
Anti-Speedy/Ringo Rabbit Polyclonal Antibody (DyLight® 550)
Antibodies
Rabbit Polyclonal Speedy/Ringo Antibody [DyLight 550]. Tested Applications: Western Blot. Tested Reactivity: Mouse.

Type: Primary
Antigen: Speedy/Ringo
Clonality: Polyclonal
Clone:
Conjugation: DyLight 550
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR